EDTA (ethylenediamine tetraacetic acid) 0.1 mol/l in aqueous solution
Catalog # AMBI9912
Supplier: Ambion
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 mL
- Seq:2370
Specifications
About this item
:
:292.25 g/mol
Storage Temperature:Ambient
EINECS:200-449-4
CAS Number:60-00-4
Formula:C₁₀H₁₆N₂O₈
MDL Number:MFCD00003541