Order Entry
Norway
ContactUsLinkComponent
Potassium chloride 2 mol/l (2 N) in aqueous solution
Potassium chloride 2 mol/l (2 N) in aqueous solution
Katalog # AMBI9640G
Supplier:  Ambion
CAS Number:  
Potassium chloride 2 mol/l (2 N) in aqueous solution
Katalog # AMBI9640G
Supplier:  Ambion
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :74.55 g/mol
Density:1.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360