Phenol red 0.04% in water for pH determination
TCIAP0101-500ML
: TCI
:
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
- Appearance:Light red to Yellow to Red clear liquid
- Transition interval (pH):(yellow)6.8 to 8.4 (red)
:
:14,07247
:354.38 g/mol
:205-609-7
:143-74-8
:C₁₉H₁₄O₅S
:MFCD00003552