Order Entry
Norway
ContactUsLinkComponent
D-Gluconic acid 45 - 50% in water (contains Gluconolactone)
Corrosive
D-Gluconic acid 45 - 50% in water (contains Gluconolactone)
Supplier:  TCI
Certificates
undefined
Corrosive
D-Gluconic acid 45 - 50% in water (contains Gluconolactone)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Red to Green clear liquid
  • Concentration (Neutralization back titration):
    45.0 to 50.0 w/w% (contains Gluconolactone)

Specification Test Results

Related Information