Order Entry
Norway
ContactUsLinkComponent
Methyl-3-amino-4-methylbenzoate ≥98.0% (by GC)
Methyl-3-amino-4-methylbenzoate ≥98.0% (by GC)
Katalog # TCIAA1432-25G
Supplier:  TCI
CAS Number:  
undefined
Methyl-3-amino-4-methylbenzoate ≥98.0% (by GC)
Katalog # TCIAA1432-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Gray to Brown powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    114.0 to 117.0 deg-C

Specification Test Results

About this item

 :
 :165.19 g/mol
 :GHS07
CAS Number:18595-18-1
Formula:C₉H₁₁NO₂
MDL Number:MFCD00025206