Sulphaquinoxaline
Catalog # EHERC16990000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:300.3357 g/mol
EINECS:200-423-2
CAS Number:59-40-5
Formula:C₁₄H₁₂N₄O₂S
MDL Number:MFCD00055406