Hood, 446
Catalog # MMMA446
Supplier: 3M
Specifications
- Garment application:Laboratory
- Colour:White
- Model:446
- Pk:300
Specifications
About this item
This hood is made of the same microporous laminated PP/PE fabric used in the 3M™ overall 4510. The hood offers a general barrier to non hazardous substances, with drapes over the shoulders offering excellent coverage.
- Low-linting fabric
- Anti-static treated
- Serged seams
- Elasticated around the face
Cat. I
Certifications: Cat. I