Order Entry
Norway
ContactUsLinkComponent
Cold resistant gloves, Cama Iso® 690+
Cold resistant gloves, Cama Iso® 690+
Supplier:  KCL by Honeywell
Certificates
undefined
Cold resistant gloves, Cama Iso® 690+
Supplier:  KCL by Honeywell
Specifications
  • Catalog No:
  • 112-1945
  • 112-1946
  • Model:
  • 690+
  • 690+
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Liner material:
  • Fleece
  • Fleece
  • Finish:
  • Rough
  • Rough
  • Cuff style:
  • Gauntlet
  • Gauntlet
  • EN norm:
  • EN 388:2016 (3221X), EN 511 (021)
  • EN 388:2016 (3221X), EN 511 (021)
Specifications