Maxi vertical dual plate electrophoresis units
Supplier: Roth Carl
Certificates
About this item
Specifications
- Catalog No:
- ROTHX951.1
- ROTHX950.1
- Model:
- Cooled
- Standard
- Glass plate size (W×H):
- 200×200 mm
- 200×200 mm
- Humidity:
- <80 % RH
- <80 % RH
- Max. power requirement:
- 1000 V
- 1000 V
- Max. temperature:
- 65 °C
- 65 °C
- Spacers:
- 1,0 mm
- 1,0 mm
- Sample capacity:
- 48
- 48
- Running buffer volume:
- 650 ml (upper tank), 1000 ml (lower tank)
- 650 ml (upper tank), 1000 ml (lower tank)