Combined metal electrodes, Radiometer Analytical
Supplier: Hach
Certificates
About this item
The Radiometer Analytical electrode range features single, double, and combined metal electrodes with platinum, silver, gold, antimony, mercury and glassy carbon sensing elements. This variety of designs can be used for redox measurements, redox titrations, potentiometric techniques, and imposed current potentiometric titration.
Ordering information: The screw cap electrodes require the CL114 cable for use with Hach sensION+ benchtop and other BNC-compatible meters/titrators.