4'-Formylbenzo-15-crown-5 (4'-formylbenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥98.0% (by GC)
: TCI
:60835-73-6
:
:C₁₅H₂₀O₆
:MFCD00192180
:296.32 g/mol
:GHS07
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:83.0 to 87.0 deg-C