2-Butynoic acid methyl ester ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:23326-27-4
:
Formula:C₅H₆O₂
MDL Number:MFCD00009273
:98.1 g/mol
Boiling Pt:148°C
Density:0,98 g/cm³ (20 °C)
Flash Pt:45°C
UN:3272
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %