Boron tribromide 1 M in dichloromethane, AcroSeal®
Catalog # 427101000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Clear yellow to brown Solution (May develop some turbidity or precipitate. May darken on storage)
- Strength:0.95 to 1.10 M
- Titration Complexometric:≥97.5 %
Specification Test Results
About this item
:
:250.52 g/mol
Density:1.467 g/cm³
Storage Temperature:Ambient
EINECS:233-657-9
UN:3390
ADR:6.1,
:GHS06
:GHS08
:GHS05
CAS Number:10294-33-4
Formula:BBr₃
MDL Number:MFCD00011312