Order Entry
Norway
Orders LinkContactUsLinkComponent
Ginkgolide B
Ginkgolide B
Certificates
Ginkgolide B

2 Options Available Below

Product Details & Documents

CAS No.: 15291-77-7 Molecular Formula: C20H24O10 Formula Weight: 424.40 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White Solubility: Soluble in DMSO. Slightly soluble in water and ethanol Merck Reference: 14,4426 MDL No.: MFCD05662347

Platelet-Activating Factor (PAF) antagonist

  • CAS Number:15291-77-7
  • Formula:C₂₀H₂₄O₁₀
  • MDL Number:MFCD05662347
  • Molecular Weight:424.40 g/mol
  • Merck Index:14,4426
Product Family Options

Product Information