Order Entry
Norway
ContactUsLinkComponent
Calcium chloride dihydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Irritant
Calcium chloride dihydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
Irritant
Calcium chloride dihydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    min. 99.0 %
  • Ammonium (NH4):
    max. 0.005 %
  • Barium (Ba):
    max. 0.003 %
  • Potassium (K):
    max. 0.01 %
  • Magnesium (Mg):
    max. 0.05 %
  • Sodium (Na):
    max. 0.01 %
  • Strontium (Sr):
    max. 0.05 %
  • Insoluble in Water:
    max. 0.005 %
  • Oxidizing Substances (as NO3):
    max. 0.003 %
  • pH of 5% Solution at 20°C:
    4.5 - 6.5
  • Sulfate (SO4):
    max. 0.005 %
  • Iron (Fe):
    max. 5
  • Heavy Metals (as Pb):
    max. 5

Specification Test Results

Related Information