Order Entry
Norway
ContactUsLinkComponent
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) ≥98.0% (by HPLC, total nitrogen) for biochemical research
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) ≥98.0% (by HPLC, total nitrogen) for biochemical research
Supplier:  TCI
Certificates
undefined
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) ≥98.0% (by HPLC, total nitrogen) for biochemical research
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Specific rotation [a]20/D:
    -60.0 to -72.0 deg (C=1, DMF)
  • beta-Galactosidase detection test:
    to pass test

Specification Test Results

Related Information