

(Formylmethyl)triphenylphosphonium chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:62942-43-2
:
Formula:C₂₀H₁₈ClOP
MDL Number:MFCD00012003
:340.79 g/mol
EINECS:263-767-2
:GHS05
:GHS07
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Methanol:within almost transparency