
4'-Methoxy-2-chloroacetophenone 97%
Katalog # ACRO425260010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:98 to 100 °C
- GC:≥96.0 %
Specification Test Results
About this item
:
:184.6196 g/mol
Melting Pt:98…100 °C
Storage Temperature:Ambient
UN:1759
ADR:8,II
:GHS05
CAS Number:2196-99-8
Formula:C₉H₉ClO₂
MDL Number:MFCD00216508