6-Aminonicotinamide, pale yellow crystalline powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:329-89-5
:
Formula:C₆H₇N₃O
MDL Number:MFCD00006327
:137.14 g/mol
EINECS:206-349-7
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
- Melting Point:240 - 250 °C