Diphenyl phosphate ≥99.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:838-85-7
:
Formula:C₁₂H₁₁O₄P
MDL Number:MFCD00003033
:250.19 g/mol
EINECS:212-657-2
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Neutralization titration):min. 99.0 %
- Melting point:67.0 to 71.0 deg-C