Order Entry
Norway
Orders LinkContactUsLinkComponent
Diphenyl phosphate ≥97%

Diphenyl phosphate ≥97%

Diphenyl phosphate ≥97%
Certificates
Diphenyl phosphate ≥97%
Diphenyl phosphate ≥97%

3 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 838-85-7 Molecular Formula: C12H11O4P Formula Weight: 250.19 Melting Point: 62-70° Beilstein Reference: 1379164

Synonyms:  Diphenyl hydrogen phosphate
  • CAS Number:838-85-7
  • Formula:C₁₂H₁₁O₄P
  • MDL Number:MFCD00003033
  • Molecular Weight:250.19 g/mol
  • Melting Pt:62…70 °C
  • Storage Temperature:Ambient
  • EINECS:212-657-2
Product Family Options

Product Information