
N-(4-Methoxybenzylidene)-4-butylaniline ≥98.0% (by GC, titration analysis)
TCIAM0275-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White or Colorless to Light orange to Yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
:
:267.3654 g/mol
:247-527-4
:GHS07
:26227-73-6
:C₁₈H₂₁NO
:MFCD00009464