
N-(4-Ethoxybenzylidene)-4-butylaniline ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:29743-08-6
:
Formula:C₁₉H₂₃NO
MDL Number:MFCD00009465
EINECS:249-821-8
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 99.0 %
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together