Order Entry
Norway
ContactUsLinkComponent
4-(Phenylazo)diphenylamine ≥97%, Sigma-Aldrich®
Irritant
4-(Phenylazo)diphenylamine ≥97%, Sigma-Aldrich®
Certificates
undefined
Irritant
4-(Phenylazo)diphenylamine ≥97%, Sigma-Aldrich®

Specification Test Results

  • Appearance (Color):
    Orange to Brown
  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Carbon:
    76.3 - 81.9 %
  • Nitrogen:
    14.8 - 15.9 %
  • Purity (HPLC):
    ≥96.5 %

Specification Test Results