

2-Nitroaniline hydrochloride ≥98.0% (by HPLC, titration analysis)
TCIAN0121-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
- Solubility in Methanol:almost transparency
:
:6.1,II
:GHS07
:GHS08
:15873-52-6
:C₆H₇ClN₂O₂
:MFCD00060216