Order Entry
Norway
ContactUsLinkComponent
(±)-Taxifolin, hydrate ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
(±)-Taxifolin, hydrate ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 580553-25
CAS Number:  
(±)-Taxifolin, hydrate ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 580553-25
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :304.25 g/mol (anhy)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
CAS Number:24198-97-8
 :480-18-2
Formula:C₁₅H₁₂O₇
MDL Number:MFCD11044454