Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
4-Methoxy-3,3'-dimethylbenzophenone
4-Methoxy-3,3'-dimethylbenzophenone
4-Methoxy-3,3'-dimethylbenzophenone
Print
4-Methoxy-3,3'-dimethylbenzophenone
Catalog #
4-Methoxy-3,3'-dimethylbenzophenone
Catalog #
Supplier:
CAS Number:
41295-28-7
Expand image
Return to Top
4-Methoxy-3,3'-dimethylbenzophenone
Add to cart
CAS Number:
41295-28-7
Return Policy
Product Details & Documents
CAS Number:
41295-28-7
Formula:
C₁₆H₁₆O₂
MDL Number:
MFCD00128925
EINECS:
255-300-6