Order Entry
Norway
ContactUsLinkComponent
(S)-(+)-3-Amino-1-boc-piperidine ≥95.0% (by GC, titration analysis)
(S)-(+)-3-Amino-1-boc-piperidine ≥95.0% (by GC, titration analysis)
  TCIAA2335-1G
 :  TCI
 :  
undefined
(S)-(+)-3-Amino-1-boc-piperidine ≥95.0% (by GC, titration analysis)
  TCIAA2335-1G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    Light yellow to Yellow to Orange clear liquid
  • Purity (GC):
    min. 95.0 %
  • Purity (Nonaqueous Titration):
    min. 95.0 %
  • Specific rotation [a]20/D:
    +27.0 to +34.0 deg (C=1, DMF)

 

 

 :
 :200.28 g/mol
 :122°C
 :GHS07
 :625471-18-3
 :C₁₀H₂₀N₂O₂
 :MFCD03094718