Order Entry
Norway
ContactUsLinkComponent
Folic acid
Folic acid
Katalog # EHERC13888000
Supplier:  EHRENSTORFER
CAS Number:  
Folic acid
Katalog # EHERC13888000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :441.41 g/mol
EINECS:200-419-0
CAS Number:59-30-3
Formula:C₁₉H₁₉N₇O₆
MDL Number:MFCD00079305