N(α)-Boc-L-histidine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:17791-52-5
:
Formula:C₁₁H₁₇N₃O₄
MDL Number:MFCD00065576
:255.3 g/mol
EINECS:241-768-9
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Specific rotation [a]20/D:+24.0 to +27.0 deg (C=1, MeOH)
- Solubility in Methanol:almost transparency