Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N(α)-Boc-L-histidine
N(α)-Boc-L-histidine
N(α)-Boc-L-histidine
N(α)-Boc-L-histidine
Supplier:
Novabiochem (Part of Merck)
Certificates
Expand image
N(α)-Boc-L-histidine
Supplier:
Novabiochem (Part of Merck)
About this item
CAS Number:
17791-52-5
:
Formula:
C₁₁H₁₇N₃O₄
MDL Number:
MFCD00065576
:
255.3 g/mol
EINECS:
241-768-9