1,1-Diphenyl-2-thiourea ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD0918-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in hot Methanol:almost transparency
Specification Test Results
About this item
:
:228.3100 g/mol
EINECS:223-448-0
CAS Number:3898-08-6
Formula:C₁₃H₁₂N₂S
MDL Number:MFCD00059151