Ficoll® PM70
Supplier: Cytiva
Certificates
About this item
Ficoll® PM70 is a high molecular weight sucrose polymer formed by copolymerisation of sucrose with epichlorohydrin. The molecules are highly branched and the high content of hydroxyl groups leads to very good solubility in aqueous media.
- Displays minimal toxicity
- Favourable viscosity and osmotic properties
The molecules are highly branched and the high content of hydroxyl groups leads to very good solubility in aqueous media.
Related Information
Certificates
Search by Lot Number17-0310-50_25 DF 17031010_DATA FILE litdoc18112350AB 20110830182032 litdoc18115827AB 20110830230116
Datasheet
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements