PIG® Absorbent pans
Supplier: NEW PIG
Certificates
About this item
Trap annoying drips in this spillproof, absorbent-filled Drip Pan. Polypropylene cover prevents splashes and keeps filler from spilling out.
- Low-profile drip pan fits underneath drum taps and other tight spaces; wide base won't tip over like unsturdy coffee cans
- Soft polypropylene cover catches drips without splashing and prevents highly absorbent polypropylene filler from spilling out
- Highly absorbent polypropylene filler soaks up liquid on contact
- Ideal for use under leaking drum taps, spigots and flanges to absorb nuisance drips; keeps your floors clean and safe
- Absorbs and retains oils, coolants, solvents and water
98% pre-consumer recycled polypropylene filler.