Order Entry
Norway
ContactUsLinkComponent
Affinity chromatography columns, MBPTrap™ HP
Affinity chromatography columns, MBPTrap™ HP
Supplier:  Cytiva
Certificates
undefined
Affinity chromatography columns, MBPTrap™ HP
Supplier:  Cytiva
Specifications
  • Catalog No:
  • 29-0486-41
  • 28-9187-78
  • 28-9187-79
  • 28-9187-80
  • Max. pressure:
  • 5 bar
  • 5 bar
  • 5 bar
  • 5 bar
  • Storage:
  • 2 to 8 °C, 20% Ethanol
  • 2 to 8 °C, 20% Ethanol
  • 2 to 8 °C, 20% Ethanol
  • 2 to 8 °C, 20% Ethanol