Sephadex® G-25 Grade, illustra™
17-0572-02
: Cytiva
:
- Pk:100 g
- Method / Format:Gel filtration
- Prep size:Scalable
- Refine target molecule:Amplified/modified DNA
- Sample type:PCR and other enzymatic reactions
- Target molecule:DNA
- Description:illustra™ Sephadex® G-25 DNA Grade SF
- Description:illustra™ Sephadex® G-25 DNA Grade SF
For purification of DNA (10 bases in length) from small molecules by gel filtration.
- Tested to ensure reproducibly high recovery of DNA
- Used in MicroSpin G-25 columns and NAP columns
- Enables self preparation of columns for nucleic acid purification