Aluminium oxide neutral, powder, BAKER ANALYZED®, J.T.Baker®
Catalog # 0540-01
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:7.0 - 7.8
- pH of 5% Slurry at 20°C:6.0 - 7.5
- Adsorption, mg o-nitroaniline/g:≥1.0
Specification Test Results
About this item
:
:101.96 g/mol
Merck Index:13,00355
Boiling Pt:2980 °C (1013 hPa)
Melting Pt:2045 °C
Density:3.94 g/cm³ (20 °C)
:Management of Change (MOC) category = N
EINECS:215-691-6
REACH:01-2119529248-35
:GHS07
:GHS08
CAS Number:1344-28-1
Formula:Al₂O₃
MDL Number:MFCD00003424