D-(+)-Raffinose pentahydrate ≥98.0% (by HPLC)
Katalog # TCIAR0002-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:78.0 to 82.0 deg-C
- Specific rotation [a]20/D:+100 to +110 deg (C=10, H2O)
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
:594.52 g/mol
Merck Index:14,08096
Melting Pt:80 °C
Density:1,516 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-146-9
CAS Number:17629-30-0
Formula:C₁₈H₃₂O₁₆·5H₂O
MDL Number:MFCD00071590