Temperature loggers, TomKey®
Supplier: bioMérieux
Certificates
About this item
The TomKey® temperature loggers are specifically designed for temperature monitoring. They are suitable for use in thermo-sensitive products, cold chain contro and transport condition records.
- Multi-function LCD with minimum/maximum alarms display
- USB 2.0 interface for quick and easy communication with the software
- Logging activation is either programmable or with the included magnet key
Delivery: Supplied with free registration and data processing software, and a calibration certificate.