TAPSO (2-hydroxy-3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid) ≥99.0% (by titrimetric analysis) Good's buffer component for biological research
Catalog # TCIAT1364-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 99.0 %
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
:259.28 g/mol
EINECS:269-993-8
CAS Number:68399-81-5
Formula:C₇H₁₇NO₇S
MDL Number:MFCD00038352