TES sodium salt (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid sodium salt) ≥99.0% (by titrimetric analysis) Good's buffer component for biological research
Catalog # TCIAT0962-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 99.0 % (calcd.on anh.substance)
- Solubility in Methanol:very faint turbidity
- Water:max. 8.0 %
Specification Test Results
About this item
:
:251.24 g/mol
Storage Temperature:Ambient
CAS Number:70331-82-7
Formula:C₆H₁₄NNaO₆S
MDL Number:MFCD00065482