

1,2,4,5-tetraklor-3-nitrobensen ≥98.0% (by GC)
: TCI
:117-18-0
:
:C₆HCl₄NO₂
:MFCD00007066
:304 °C (1013 hPa)
:99 °C
:1,744 g/cm³ (25 °C)
:204-178-2
:3077
:9,III
:260.89 g/mol
:GHS07
:GHS09
:Romtemperatur
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:99.0 to 102.0 deg-C