4-(Dimetylamino)bensosyre ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:619-84-1
:
Formula:C₉H₁₁NO₂
MDL Number:MFCD00002537
:165.19 g/mol
Merck Index:14,03232
Boiling Pt:297 °C (1013 hPa)
Melting Pt:243°C
Density:1,29 g/cm³ (20 °C)
EINECS:210-615-8
:GHS07
Storage Temperature:Romtemperatur
Specification Test Results
- Appearance:White to Light yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Methanol:almost transparency