Order Entry
Norway
ContactUsLinkComponent
4-Chloro-3-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
4-Chloro-3-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier:  TCI
Certificates
undefined
4-Chloro-3-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    97.0 to 109.0 %

Specification Test Results

Related Information