
3,3',5,5'-Tetramethylbenzidine (TMB) ≥98.0% (by HPLC, titration analysis) for biochemical research
Supplier: TCI
Certificates
About this item
CAS Number:54827-17-7
:
Formula:C₁₆H₂₀N₂
MDL Number:MFCD00007748
:240.35 g/mol
Boiling Pt:402 °C (1013 hPa)
Melting Pt:166…170 °C
Storage Temperature:Ambient
EINECS:259-364-6
:GHS07
Specification Test Results
- Appearance:White to Green to Brown powder to crystaline
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:167.0 to 172.0 deg-C
- Peroxidase detection test:to pass test