L(-)-Malic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:97-67-6
:
Formula:C₄H₆O₅
MDL Number:MFCD00064213
:134.09 g/mol
Merck Index:14,05707
Boiling Pt:322 °C (1013 hPa)
Melting Pt:101…104 °C
Density:1,595 g/cm³ (20 °C)
EINECS:202-601-5
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Optical purity (GC):min. 98.0 ee%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:101.0 to 105.0 deg-C