

1,5-Dihydroxy-9,10-anthraquinone ≥85.0% (by HPLC)
Catalog # TCIAD0562-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 85.0 area%
Specification Test Results
About this item
:
:240.22 g/mol
Merck Index:14,00689
EINECS:204-175-6
:GHS07
:GHS08
CAS Number:117-12-4
Formula:C₁₄H₈O₄
MDL Number:MFCD00001210