DL-Isoleucine mixture of diasteroisomers ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:443-79-8
:
Formula:C₆H₁₃NO₂
MDL Number:MFCD00004268
:131.17 g/mol
Merck Index:14,05179
Melting Pt:275 °C
Density:1,255 g/cm³ (-123.15 °C)
EINECS:207-139-8
Specification Test Results
- Appearance:White powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Specific rotation [a]20/D:-2.0 to +2.0 deg (C=4, 6mol/L HCl)