Order Entry
Norway
ContactUsLinkComponent
 
Irritant
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)phenol ≥97.0% (by GC)
Catalog # TCIAT1983-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)phenol ≥97.0% (by GC)
Catalog # TCIAT1983-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :GHS07
CAS Number:2787-79-3
Formula:C₇HF₇O
MDL Number:MFCD00155984