Order Entry
Norway
ContactUsLinkComponent
Ethyl phenylcyanoacetate 97%
Ethyl phenylcyanoacetate 97%
Catalog # ACRO118830050
CAS Number:  
undefined
Ethyl phenylcyanoacetate 97%
Catalog # ACRO118830050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5033 to 1.5053 (20°C, 589 nm)
  • Titration with NaOH:
    ≥96.0 %

Specification Test Results

About this item

 :
Density:1.09 g/cm³
Storage Temperature:Ambient
EINECS:224-921-4
ADR:6.1,II
 :189.21 g/mol
UN:2810
 :GHS07
CAS Number:4553-07-5
Formula:C₁₁H₁₁NO₂
MDL Number:MFCD00001866